missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoamyl Alcohol, (Isopentyl Alcohol), Reagents
Specifications
Specifications
| Boiling Range | 131°C to 132.5°C |
| Melting Point | -117.2°C |
| Color | Colorless |
| Boiling Point | 131°C to 132.5°C |
| Melting Range | -117.2°C |
| Packaging | Glass Bottle |
| Quantity | 1 L |
| Specific Gravity | 0.813 |
| Grade | Reagent |
| Physical Form | Liquid |
| Show More |