missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Alcohol, (Isopentyl Alcohol), Reagents

Catalog No. 01337555
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337555 1 L
01337554 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337555 Supplier Labchem Inc Supplier No. C21195001C
Only null left

Isoamyl Alcohol, (Isopentyl Alcohol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym Isopentyl alcohol
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Boiling Range 131°C to 132.5°C
Melting Point -117.2°C
Color Colorless
Boiling Point 131°C to 132.5°C
Melting Range -117.2°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 0.813
Grade Reagent
Physical Form Liquid
Chemical Name or Material Isoamyl Alcohol, (Isopentyl Alcohol)
Show More Show Less