missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Acrylate (stabilized with HQ) 98.0+%, TCI America™

Catalog No. A089025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A089025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A089025ML Supplier TCI America Supplier No. A089025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4245-35-6
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00053725
InChI Key ZVYGIPWYVVJFRW-UHFFFAOYSA-N
Synonym Acrylic Acid Isoamyl Ester, Isopentyl Acrylate, Acrylic Acid Isopentyl Ester
PubChem CID 77920
IUPAC Name 3-methylbutyl prop-2-enoate
SMILES CC(C)CCOC(=O)C=C

Specifications

Color Colorless
Boiling Point 159°C
Quantity 25 mL
UN Number 3272
Formula Weight 142.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Acrylate (stabilized with HQ)
EINECSNumber (2)-3442
TSCA No