missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl 4-Methoxycinnamate 95.0+%, TCI America™

Catalog No. I08155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08155G 5 g
I081525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08155G Supplier TCI America Supplier No. I08155G
Only null left

Chemical Identifiers

CAS 71617-10-2
Molecular Formula C15H20O3
Molecular Weight (g/mol) 248.322
MDL Number MFCD00583856
InChI Key UBNYRXMKIIGMKK-RMKNXTFCSA-N
Synonym 4-Methoxycinnamic Acid Isoamyl Ester, Isopentyl 4-Methoxycinnamate, 4-Methoxycinnamic Acid Isopentyl Ester
PubChem CID 1549789
IUPAC Name 3-methylbutyl (E)-3-(4-methoxyphenyl)prop-2-enoate
SMILES CC(C)CCOC(=O)C=CC1=CC=C(C=C1)OC

Specifications

Color Yellow
Boiling Point 158°C
Quantity 5 g
Formula Weight 248.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl 4-Methoxycinnamate
RTECSNumber UD3393500
TSCA No