Learn More
GA International ISO Warning Labels - ""Hot Surface Hazard Symbol"" 0.75"" x 0.75"" / 19.1mm x 19.1mm Rectangle Black & Yellow on Silver From -80C to +100C (-112F to +212F)
Catalog No. 5000572280
Encompass
Pre-printed, hot surface burn hazard warning labels for the identification of equipment and material that can pose a risk due to a heated surface. Provided with a glove-friendly adhesive, these waterproof thermoplastic, high-heat labels are perfect for labeling water baths, heat blocks, autoclave containers, and incubators.
Catalog No. 5000572280 Supplier GA International Supplier No. WL0080.25R
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Pre-printed, hot surface burn hazard warning labels for the identification of equipment and material that can pose a risk due to a heated surface. Provided with a glove-friendly adhesive, these waterproof thermoplastic, high-heat labels are perfect for labeling water baths, heat blocks, autoclave containers, and incubators.