missing translation for 'onlineSavingsMsg'
Learn More

Iso-Propyl Alcohol/Acetic Acid Solution, Reagents

Catalog No. 01334928
Change view
Click to view available options
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01334928 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334928 Supplier Labchem Inc Supplier No. CI171900500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Iso-Propyl Alcohol/Acetic Acid Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Iso-Propyl Alcohol/Acetic Acid Solution
CAS 7732-18-5, 67-63-0
Color Colorless
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.8913
Grade Reagent