missing translation for 'onlineSavingsMsg'
Learn More

Ismatec™ Extension Pump Tubing, PharMed™ BPT

Ismatec™ Extension Pump Tubing protects light-sensitive samples

$1018.82 - $1358.44

Specifications

Autoclavable Autoclavable
Material PharMed BPT
Overall Length (English) 100 ft.
For Use With (Application) Ismatec pumps
Color Tan
View More Specs

Products 8
Catalog Number Mfr. No. Tubing Size Price Qty  
Catalog Number Mfr. No. Tubing Size Price Qty  
13200073
View Documents
Ismatec™
MFLX9580912
0.25 mm x 30.4 m L
Each for $1,018.82
Only null left
 
13-200-074
View Documents
Ismatec™
MFLX9580914
0.38 mm x 30.4 m L
Each for $1,018.82
Only null left
 
13200075
View Documents
Ismatec™
MFLX9580926
0.89 mm x 30.4 m L
Each for $1,358.44
Only null left
 
13-200-076
View Documents
Ismatec™
MFLX9580928
1.02 mm x 30.4 m L
Each for $1,018.82
Only null left
 
13200077
View Documents
Ismatec™
MFLX9580930
1.14 mm x 30.4 m L
Each for $1,358.44
Only null left
 
13200078
View Documents
Ismatec™
MFLX9580934
1.42 mm x 30.4 m L
Each for $1,018.82
Only null left
 
13200079
View Documents
Ismatec™
MFLX9580942
2.06 mm x 30.4 m L
Each for $1,018.82
Only null left
 
13200080
View Documents
Ismatec™
MFLX9580948
2.79 mm x 30.4 m L
Each for $1,018.82
Only null left
 
Description

Description

  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Specifications

    Specifications

    Autoclavable
    100 ft.
    Tan
    PharMed BPT
    Ismatec pumps
    Extension Pump Tubing
    Videos
    SDS
    Documents

    Documents