missing translation for 'onlineSavingsMsg'
Learn More

Ishikawa's Reagent 92.0+%, TCI America™

Catalog No. H059825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H059825G 25 g
H0598100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H059825G Supplier TCI America Supplier No. H059825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 309-88-6
Molecular Formula C7H11F6N
Molecular Weight (g/mol) 223.162
MDL Number MFCD00060264
InChI Key BNTFCVMJHBNJAR-UHFFFAOYSA-N
Synonym n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant
PubChem CID 136149
IUPAC Name N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine
SMILES CCN(CC)C(C(C(F)(F)F)F)(F)F

Specifications

Color Yellow
Boiling Point 54°C
Quantity 25 g
UN Number 2920
Formula Weight 223.16
Percent Purity ≥92.0% (T)
Physical Form Liquid
Chemical Name or Material Ishikawa′s Reagent [Fluorinating Reagent]