Learn More
YSI™ ISE Probes for Pro Series Portable Meters
Field probes for ion concentration measurements.
$1190.04 - $1268.91
Specifications
| Applications | For use in fresh water applications |
|---|---|
| For Use With (Equipment) | YSI Pro Series Portable Meters |
| Catalog Number | Mfr. No. | Parameter | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Parameter | Price | Quantity | |||||
|
15-177-463
|
YSI™
YSI605104 |
Ammonium |
Each for $1,190.04
|
|
|||||
|
15-177-464
|
YSI™
605105 |
Chloride |
Each for $1,268.91
|
|
|||||
|
15-177-465
|
YSI™
605106 |
Nitrate |
Each for $1,190.04
|
|
|||||
Description
For use with Pro Series Portable Meters.Specifications
| For use in fresh water applications | |
| YSI Pro Series Portable Meters |