missing translation for 'onlineSavingsMsg'
Learn More

Isatoic Anhydride 98.0+%, TCI America™

Catalog No. I008125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I008125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I008125G Supplier TCI America Supplier No. I008125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 118-48-9
Color White
Molecular Formula C8H5NO3
Quantity 25 g
MDL Number MFCD00006700
Synonym isatoic anhydride, 2h-3,1-benzoxazine-2,4 1h-dione, 1h-benzo d 1,3 oxazine-2,4-dione, isatoic acid anhydride, 4h-3,1-benzoxazine-2,4 1h-dione, isato acid anhydride, n-carboxyanthranilic anhydride, unii-r8tfa74y4u, 3,1-benzoxazine-2,4 1h-dione, anthranilic acid, n-carboxy-, cyclic anhydride
InChI Key TXJUTRJFNRYTHH-UHFFFAOYSA-N
SMILES O=C1NC2=CC=CC=C2C(=O)O1
IUPAC Name 2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
Molecular Weight (g/mol) 163.13
PubChem CID 8359
Formula Weight 163.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isatoic Anhydride
Show More Show Less
EINECSNumber (5)-0856
RTECSNumber DM3100000
TSCA Yes