missing translation for 'onlineSavingsMsg'
Learn More

Isatin 98.0+%, TCI America™

Catalog No. I008025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I008025G 25 g
I0080100G 100 g
I0080500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I008025G Supplier TCI America Supplier No. I008025G

Chemical Identifiers

CAS 91-56-5
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.133
MDL Number MFCD00005718
InChI Key JXDYKVIHCLTXOP-UHFFFAOYSA-N
Synonym isatin, indoline-2,3-dione, 2,3-indolinedione, indole-2,3-dione, isatine, 2,3-dioxoindoline, pseudoisatin, tribulin, isotin, isatic acid lactam
PubChem CID 7054
ChEBI CHEBI:27539
IUPAC Name 1H-indole-2,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(=O)N2

Specifications

Melting Point 203°C
Quantity 25 g
Formula Weight 147.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isatin
EINECSNumber (5)-0148
RTECSNumber NL7873000
TSCA Yes