missing translation for 'onlineSavingsMsg'
Learn More

ISA (ionic strength adjustment Solution: 1 M KCl) 1M KCl, MilliporeSigma™ Supelco™

Catalog No. 111015419
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015419 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015419 Supplier Merck Emd Millipore Supplier No. 58221500MLF
Only null left

Specifications

CAS 7447-40-7
Linear Formula KCl
Concentration 1 M KCl
MDL Number MFCD00011360
Synonym Potassium chloride solution
Type Organic Standard
Quantity 500 mL
Formula Weight 74.55