Learn More
Air Boss™ Defense IRT Water Storage Bladders
Effectively collect wastewater runoff from IRT decontamination shelters
$6,750.93
Specifications
| Product Type | Waste Storage Bladder |
|---|---|
| Material | Polyester |
| For Use With (Application) | Collects waste water run-off from IRT decontamination shelters |
| Includes | Protective ground Cloth and transport bag, integral pressure relief valve, ball valve, back flow prevention valve and cam-lok |
Includes: Protective ground cloth and transport bag
| Catalog Number | Mfr. No. | Capacity (Metric) | Capacity (English) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Capacity (English) | Price | Quantity & Availability | ||||
191300635
|
Air Boss™ Defense
WSB250G |
346.3 L | 250 gal. |
Each for $6,750.93
|
|
||||
Description
- Co-polymer material is highly resistant to chemicals, mold and mildew
- Integral pressure relief valve, ball valve, back flow prevention valve, and cam-lok
- Ball valve on either side of cam-lok for shut off, backflow prevention for one way fluid exchange, cam-lok for quick disconnect
- Rugged design allows for use in the toughest environments
- Quickly and easily deploys, no tools required, reusable and repairable
- Compact and lightweight for portability, fully collapsible and foldable
Specifications
| Waste Storage Bladder | |
| Polyester | |
| Collects waste water run-off from IRT decontamination shelters | |
| Protective ground Cloth and transport bag, integral pressure relief valve, ball valve, back flow prevention valve and cam-lok |