Learn More
SignalChem Biotech IGF1R, Active 50ug
Catalog No. 502574669
Encompass
Recombinant human IGF1R (960-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 502574669 Supplier SignalChem Biotech Supplier No. I0211H50
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human IGF1R (960-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.