missing translation for 'onlineSavingsMsg'
Learn More

Iron wire, 2.0mm (0.08in) dia, Puratronic™, 99.995% (metals basis)

Form: Approx.25g/m | CAS: 7439-89-6 | Fe | 55.85 g/mol

$599.32 - $1785.12

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10934BU
View Documents
Thermo Scientific Chemicals
010934BU
25 cm
Each for $599.32
Only null left
 
AA10934BY
View Documents
Thermo Scientific Chemicals
010934BY
100 cm
Each for $1,785.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-89-6
55.85
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
CHEBI:82664
[Fe]

Specifications

Gray
Gray wire
99.995% (Metals basis)
Fe
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
[Fe]
55.85
CHEBI:82664
7439-89-6
25 cm
Wire
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
Iron
Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 231-096-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only