missing translation for 'onlineSavingsMsg'
Learn More

Iron wire, 0.5mm (0.02in) dia, 99.99% (metals basis)

Catalog No. p-4412907
Change view
Click to view available options
Quantity:
1 m
5 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40498G1 1 m
AA40498G5 5 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40498G1 Supplier Thermo Scientific Chemicals Supplier No. 040498G1
Only null left

CAS: 7439-89-6 | Fe | 55.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Quantity 1 m
Assay Percent Range 99.99% (metals basis)
Chemical Name or Material Iron wire
Hazard Category H500
EINECSNumber 231-096-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only