missing translation for 'onlineSavingsMsg'
Learn More

Iron powder, spherical, 1-3 micron, 98+%, Thermo Scientific Chemicals

Catalog No. AA4033795
Change view
Click to view available options
Quantity:
100 g
2 x 500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4033795 2 x 500 g
AA4033722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4033795 Supplier Thermo Scientific Chemicals Supplier No. 04033795

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Carbonyl iron powder

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Color Gray
Appearance Gray powder
Quantity 2 x 500 g
Form Powder
Assay Percent Range >98% (Metals basis)
Chemical Name or Material Iron
Hazard Category H500-X
Precautionary Statement P235+P410-P280
DOTInformation DOT Class: 4.2; Packaging Group: II
EINECSNumber 231-096-4
RTECSNumber NO4565500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only