missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Iron powder, -20 mesh, 99.9% (metals basis)
Specifications
Specifications
| Melting Point | 1538° |
| CAS | 7439-89-6 |
| Boiling Point | 2740° |
| Quantity | 50 g |
| Form | Solid |
| MDL Number | MFCD00010999 |
| UN Number | UN3089 |
| Percent Purity | 99.9% |