missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Iron powder, -20 mesh, 99.9% (metals basis)

Catalog No. AA4735518
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4735518 50 g
AA4735530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4735518 Supplier Thermo Scientific™ Supplier No. 4735518

Specifications

Melting Point 1538°
CAS 7439-89-6
Boiling Point 2740°
Quantity 50 g
Form Solid
MDL Number MFCD00010999
UN Number UN3089
Percent Purity 99.9%