missing translation for 'onlineSavingsMsg'
Learn More

Iron pieces, irregularly shaped, 25mm (1.0in) & down, 99.97+% (metals basis)

Catalog No. AA10213A1
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10213A1 1 kg
AA10213A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA10213A1 Supplier Thermo Scientific Chemicals Supplier No. 010213A1

Form: 90% 2.5-25mm (0.1-1.0 in.) | CAS: 7439-89-6 | Fe | 55.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Color Gray
Appearance Gray irregularly shaped pieces
Quantity 1 kg
Form Irregular Pieces
Assay Percent Range >99.97% (Metals basis)
Formula Weight 55.847
Chemical Name or Material Iron
Hazard Category H500
EINECSNumber 231-096-4
RTECSNumber NO4565500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only