missing translation for 'onlineSavingsMsg'
Learn More

Iron phosphide, 99.5% (metals basis)

Catalog No. p-4781945
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4348609 10 g
AA4348618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4348609 Supplier Thermo Scientific Chemicals Supplier No. 04348609
Only null left

CAS: 12023-53-9 | Fe3P | 198.51 g/mol

Iron phosphide is used as a semiconductor. It finds application in high power and high frequency applications like laser diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12023-53-9
Molecular Formula Fe3P
Molecular Weight (g/mol) 198.51
MDL Number MFCD10567221
InChI Key KFSKMQQNHSLZRJ-UHFFFAOYSA-N
IUPAC Name triferriophosphane
SMILES [Fe]P([Fe])[Fe]

Specifications

Color Gray
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.5% (Metals basis)
Solubility Information Soluble in nitric acid,hydrogen fluoride and aqua regia. Insoluble in water,dilute acid and alkalis.
Formula Weight 198.51
Density 6.74g/cm3 at 20°C
Chemical Name or Material Iron phosphide
EINECSNumber 234-682-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only