missing translation for 'onlineSavingsMsg'
Learn More

Iron Metal, Reduced, Powder, 96%, Spectrum™ Chemical

Catalog No. 18611090
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611090 125 g Poly Bottle
18611092 500 g Poly Bottle
18611094 2.5 kg Poly Bottle
18611091 12 kg Poly Pail
18611093 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611090 Supplier Spectrum Chemical Mfg Cor Supplier No. I1035125GM

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
IUPAC Name iron
SMILES [Fe]

Specifications

CAS Max % 1
Quantity 125 g
Packaging Poly Bottle
Form Powder
Percent Purity 96%
Grade Reagent
Chemical Name or Material Iron Metal, Reduced

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.