missing translation for 'onlineSavingsMsg'
Learn More

Iron Metal, Reduced, 100 Mesh, Powder, Spectrum™ Chemical

Catalog No. p-7165737
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611086 500 g Poly Bottle
18611087 2.5 kg Poly Bottle
18611088 12 kg Poly Bottle
18611089 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611086 Supplier Spectrum Chemical Mfg Cor Supplier No. I1040500GM
Only null left

Chemical Identifiers

CAS 7439-89-6
Molecular Weight (g/mol) 55.85

Specifications

CAS Max % 1
Quantity 500 g
Packaging Poly Bottle
Form Powder
Grade Ungraded
Chemical Name or Material Iron Metal, Reduced

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.