missing translation for 'onlineSavingsMsg'
Learn More

Iron Metal, Electrolytic, Granular, Primary Standard, Approx .99.90-100%, Spectrum™ Chemical

Catalog No. p-7157620
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603509 25 g Amber Glass Bottle
18603510 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18603509 Supplier Spectrum Chemical Mfg Cor Supplier No. I119725GM
Only null left

I1197, 7439-89-6, Fe

Spectrum™ Chemical Iron Metal, Electrolytic, Granular, Primary Standard

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
IUPAC Name iron
SMILES [Fe]

Specifications

CAS Max % 100%
Quantity 25 g
Packaging Amber Glass Bottle
Form Solid
Percent Purity ∼99.9 to 100%
Grade ACS
Chemical Name or Material Iron Metal