missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron Metal, Electrolytic, Granular, Primary Standard, Approx .99.90-100%, Spectrum™ Chemical
Description
Spectrum™ Chemical Iron Metal, Electrolytic, Granular, Primary Standard
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 25 g |
| Packaging | Amber Glass Bottle |
| Form | Solid |
| Percent Purity | ∼99.9 to 100% |
| Grade | ACS |
| Chemical Name or Material | Iron Metal |