missing translation for 'onlineSavingsMsg'
Learn More

Iron Metal, Approx. 40 Mesh, Filings, Spectrum™ Chemical

$303.33 - $4934.80

Chemical Identifiers

CAS 7439-89-6
Molecular Weight (g/mol) 55.85
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611082
View Documents
Spectrum Chemical Mfg Cor
I1030500GM
500 g Poly Bottle
Each for $303.33
Only null left
 
18611083
View Documents
Spectrum Chemical Mfg Cor
I103025KG
2.5 kg Poly Bottle
Each for $1,197.63
Only null left
 
18611084
View Documents
Spectrum Chemical Mfg Cor
I103012KG
12 kg Poly Bottle
Each for $2,691.87
Only null left
 
18611085
View Documents
Spectrum Chemical Mfg Cor
I103045KGBL
45 kg Poly Bottle
Each for $4,934.80
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7439-89-6
55.85

Specifications

7439-89-6
500 g
Fine Granular Powder
Ungraded
1
Poly Bottle
55.85
Iron Metal
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.