missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron Metal, 0.25 mm Diameter, Wire, 99.99%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 50 cm |
| Packaging | Poly Bag |
| Form | Wire |
| Percent Purity | 99.99% |
| Grade | Reagent |
| Chemical Name or Material | Iron Metal |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.