missing translation for 'onlineSavingsMsg'
Learn More

Iron Metal, 0.25 mm Diameter, Wire, 99.99%, Spectrum™ Chemical

Catalog No. 18611081
Change view
Click to view available options
Quantity:
50 cm
250 cm
Packaging:
Poly Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611081 250 cm Poly Bag
18611080 50 cm Poly Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18611081 Supplier Spectrum Chemical Mfg Cor Supplier No. I1042250CM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7439-89-6
Molecular Weight (g/mol) 55.85

Specifications

CAS Max % 1
Quantity 250 cm
Packaging Poly Bag
Form Wire
Percent Purity 99.99%
Grade Reagent
Chemical Name or Material Iron Metal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.