missing translation for 'onlineSavingsMsg'
Learn More

Iron Metal, 0.25 mm Diameter, Wire, 99.99%, Spectrum™ Chemical

Catalog No. 18611080
Change view
Click to view available options
Quantity:
50 cm
250 cm
Packaging:
Poly Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611080 50 cm Poly Bag
18611081 250 cm Poly Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18611080 Supplier Spectrum Chemical Mfg Cor Supplier No. I104250CM

Chemical Identifiers

CAS 7439-89-6
Molecular Weight (g/mol) 55.85

Specifications

CAS Max % 1
Quantity 50 cm
Packaging Poly Bag
Form Wire
Percent Purity 99.99%
Grade Reagent
Chemical Name or Material Iron Metal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.