missing translation for 'onlineSavingsMsg'
Learn More

Iron lumps, vacuum remelted, low Oxygen, 99.99% (metals basis)

Catalog No. p-7049118
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
Catalog No. Quantity
AA1295814 25 g
AA1295822 100 g
2 options

Catalog No. AA1295814

Supplier: Thermo Scientific Chemicals 01295814

Only null left

CAS: 7439-89-6 | Fe | 55.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Quantity 25 g
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Iron lumps, vacuum remelted, Low Oxygen, Oxygen and Nitrogen analysis included
Hazard Category H500
EINECSNumber 231-096-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only