missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) trifluoromethanesulfonate, tech. 90%

CAS: 63295-48-7 | C3F9FeO9S3 | 503.035 g/mol

Supplier:  Thermo Scientific Chemicals H6003403

 View more versions of this product

Catalog No. AAH6003403


Only null left
Explore available promotions

Description

Description

Iron(III) triflate is used as an efficient catalyst in the synthesis of 2,3-unsaturated-O-glycosides from 2,4,6-tri-O-acetyl-D-glucal. It is also involved in the synthesis of a variety of beta-enamino ketones and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(III) triflate is used as an efficient catalyst in the synthesis of 2,3-unsaturated-O-glycosides from 2,4,6-tri-O-acetyl-D-glucal. It is also involved in the synthesis of a variety of beta-enamino ketones and esters.

Solubility
Soluble in water, methanol and acetonitrile.

Notes
Moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Iron(III) trifluoromethanesulfonate
183°C
C3F9FeO9S3
MFCD15144785
Moisture sensitive
Soluble in water,methanol and acetonitrile.
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Fe+3]
503.035
503.04
Technical
63295-48-7
110°C (230°F)
Fe(OSO2CF3)3
1 g
iron iii trifluoromethanesulfonate, fe otf 3, ferric triflate, iron iii triflate, fe cf3so3 3, ksc497q0p, iron 3+ tritriflate, ferric trifluoromethanesulfonate, iron 1,1,1-trifluoromethanesulfonic acid, iron 3+ ion tritrifluoromethanesulfonate
OSHOQERNFGVVRH-UHFFFAOYSA-K
iron(3+);trifluoromethanesulfonate
11656316
90%

Chemical Identifiers

63295-48-7
503.035
OSHOQERNFGVVRH-UHFFFAOYSA-K
11656316
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Fe+3]
C3F9FeO9S3
MFCD15144785
iron iii trifluoromethanesulfonate, fe otf 3, ferric triflate, iron iii triflate, fe cf3so3 3, ksc497q0p, iron 3+ tritriflate, ferric trifluoromethanesulfonate, iron 1,1,1-trifluoromethanesulfonic acid, iron 3+ ion tritrifluoromethanesulfonate
iron(3+);trifluoromethanesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only