missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis, Honeywell Fluka™

Catalog No. p-7121617
Change view
Click to view available options
Quantity:
500 g
1 kg
6 x 1 kg
Packaging:
Poly bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000857 500 g Poly bottle
60045925 1 kg Poly bottle
60045926 6 x 1 kg Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000857 Supplier Honeywell Inc Supplier No. 31235500G
Only null left

Puriss. p.a., 21-23% Fe basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 15244-10-7
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
MDL Number MFCD00011007
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
PubChem CID 167265
IUPAC Name iron(3+);trisulfate;hydrate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Quantity 500 g
Linear Formula Fe2(SO4)3 · xH2O
UN Number 3260
Formula Weight 399.88g/mol (anhydrous basis)
Grade puriss. p.a.
Concentration or Composition (by Analyte or Components) 21 to 23% Fe basis
Packaging Poly bottle
Chemical Name or Material Iron(Iii) Sulfate Hydrate
Hazard Statement P280-P301 + P312 + P330-P305 + P351 + P338 + P310
Precautionary Statement H290-H302-H315-H318
RTECSNumber NO8520000