missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis, Solstice

Catalog No. 60045926
Change view
Click to view available options
Quantity:
500 g
1 kg
6 x 1 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045926 6 x 1 kg Poly Bottle
6000857 500 g Poly Bottle
60045925 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60045926 Supplier Honeywell Inc Supplier No. 312356X1KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., 21-23% Fe basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 15244-10-7 , 7664-93-9
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
MDL Number MFCD00011007
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
Synonym Ferric sulfate hydrate
PubChem CID 167265
IUPAC Name iron(3+);trisulfate;hydrate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 98.0000%, 2.0000%
Quantity 6 x 1 kg
Linear Formula Fe2(SO4)3 · xH2O
UN Number UN3260
Formula Weight 399.88
Percent Purity 21 to 23% (Fe basis)
Grade puriss. p.a.
Packaging Poly Bottle
Chemical Name or Material Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis
Hazard Statement P280-P301 + P312 + P330-P305 + P351 + P338 + P310
Precautionary Statement H290-H302-H315-H318
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Corrosive solid, acidic, inorganic, n.o.s.
RTECSNumber NO8520000
Recommended Storage Room Temp