missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Iron(III) hypophosphite
Specifications
Specifications
| CAS | 7783-84-8 |
| Physical Form | Solid |
| Quantity | 100 g |
| Molecular Formula | Fe(H{2}PO{2}){3} |
| MDL Number | MFCD00049454 |
| Formula Weight | 250.81 |