missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Iron(III) hypophosphite

Catalog No. AA4165214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4165214 25 g
AA4165222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4165214 Supplier Thermo Scientific™ Supplier No. 4165214

Specifications

CAS 7783-84-8
Physical Form Solid
Quantity 25 g
Molecular Formula Fe(H{2}PO{2}){3}
MDL Number MFCD00049454
Formula Weight 250.81