missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) hydroxide, alpha-phase

Catalog No. p-4407481
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1949606 5 g
AA1949614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1949606 Supplier Thermo Scientific Chemicals Supplier No. 01949606
Only null left

For Use in Purifying Water, As Absorbent in Chemical Processes, As a Pigment | CAS: 20344-49-4 | FeHO2 | 88.851 g/mol

Iron(III) hydroxide is used in purifying water and as an absorbent in chemical processes. It is also used as a pigment and as a catalyst. Further, it is used in some tattoo ink and in aquarium water treatment as a phosphate binder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20344-49-4
Molecular Formula FeHO2
Molecular Weight (g/mol) 88.851
MDL Number MFCD00064782
InChI Key AEIXRCIKZIZYPM-UHFFFAOYSA-M
Synonym hydroxy oxo iron, goethite, lepidocrocite, iron hydroxide oxide, goethite fe oh o, iron hydroxide oxide fe oh o, iron iii oxide-hydroxide, ferric acid, iron pigment yellow, ferric hydroxide oxide
PubChem CID 91502
IUPAC Name hydroxy(oxo)iron
SMILES O[Fe]=O

Specifications

Color Brown
Physical Form Powder
Quantity 5 g
Solubility Information Soluble in mineral acids. Insoluble in water and ethanol.
Formula Weight 88.85
Appearance Brown powder
Density 3.4 to 3.9 g/cm3
Chemical Name or Material Iron(III) hydroxide
EINECSNumber 243-746-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only