missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) citrate tribasic monohydrate 18-20 Fe basis (T), 18-20% Fe basis (T), MilliporeSigma™ Supelco™

Catalog No. 111013618
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013618 250 g
111013620 1 kg
111013619 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111013618 Supplier Merck Emd Millipore Supplier No. 44941250G

Chemical Identifiers

MDL Number MFCD00013098
Synonym Ferric citrate monohydrate

Specifications

CAS 2338-05-8
Quantity 250 g
Linear Formula C6H5 FeO7 · H2O
Formula Weight 262.96
Percent Purity 18 to 20% Fe basis (T)