missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) citrate hydrate, 98%

Catalog No. AC211850010
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211850010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC211850010 Supplier Thermo Scientific Chemicals Supplier No. 211850010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 334024-15-6 | C6H5FeO7 | 244.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 334024-15-6
Molecular Formula C6H5FeO7
Molecular Weight (g/mol) 244.94
MDL Number MFCD00149615
InChI Key NPFOYSMITVOQOS-UHFFFAOYSA-K
Synonym ferric citrate monohydrate, iron iii citrate hydrate, unii-334s8t6lke, ferric citrate hydrate, ferric citrate usan, iron iii citrate tribasic monohydrate, citric acid iron iii salt, iron cento citrate hydrate, ksc534c4h, iron 3+ citrate hydrate
PubChem CID 22178220
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylate;iron(3+);hydrate
SMILES [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 18 to 22% (Fe) (Iodometry)
Packaging Plastic bottle
Quantity 1 kg
Merck Index 14, 4021
Solubility Information Solubility in water: 5g/L (25°C). Other solubilities: insoluble in ethanol, soluble in hno3
Formula Weight 262.96
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Iron(III) citrate hydrate, 98%
Show More Show Less
Hazard Category LIGHT SENSITIVE

RUO – Research Use Only