missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) 2-ethylhexanoate diisopropoxide, 10% w/v in isopropanol, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Quantity | 100 mL |
| UN Number | UN1219 |
| Sensitivity | Moisture sensitive |
| Formula Weight | 317.23 |
| Concentration or Composition (by Analyte or Components) | 10% w/v in isopropanol |
| Physical Form | Liquid |
| Chemical Name or Material | Iron(III) 2-ethylhexanoate diisopropoxide |
Safety and Handling
| Hazard Category | H225-H315-H318-H335-H336 |
| Hazard Statement | GHS H Statement H225-H361-H315-H319-H336 Highly flammable liquid and vapor. Suspected of damaging fertility or the unborn child. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL |
| TSCA | No |
| Recommended Storage | Ambient temperatures |