missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) 2,4-pentanedionate

Catalog No. p-7049026
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1253422 100 g
AA1253436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1253422 Supplier Thermo Scientific Chemicals Supplier No. 01253422

CAS: 14024-18-1 | C15H21FeO6 | 353.17 g/mol

Iron(III) 2,4-pentanedionate is used as a precatalyst and reagent in organic chemistry. It acts as a precursor for highly crystalline (Zn, Fe)Fe2O4 films and magnetic property measurements of these films. It catalyzes the dimerization of isoprene to a mixture of 1,5-dimethyl-1,5-cyclooctadiene and 2,5-dimethyl-1,5-cyclooctadiene, ring-opening polymerization of 1,3-benzoxazine and catalyze the reaction of N-sulfonyl oxaziridines with olefins to form 1,3-oxazolidine products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14024-18-1
Molecular Formula C15H21FeO6
Molecular Weight (g/mol) 353.17
MDL Number MFCD00000020
InChI Key AQBLLJNPHDIAPN-LNTINUHCSA-K
Synonym Iron(III) acetylacetonate; Tris(acetylacetonato)iron(III)
IUPAC Name iron(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point ∼183°C (decomposition)
Color Red
Quantity 100 g
Beilstein 4157960
Solubility Information Soluble in toluene.
Formula Weight 353.17
Physical Form Powder
Chemical Name or Material Iron(III) 2,4-pentanedionate
Hazard Category H302-H319-H335-H500
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
EINECSNumber 237-853-5
RTECSNumber NO8960000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only