Learn More
Iron(II) sulfide, tech.
CAS: 1317-37-9 | FeS | 87.91 g/mol
Supplier: Thermo Scientific Chemicals A155690I
Description
Iron(II) sulfide is used as a pigment in hair dyes and ceramics, glasses and bottles, and paint, Also used in lubricants and anodes, as a hydrogen sulfide source, and to treat exhaust gases. And it is also used as resulphurizing agent in the production of carbon, alloy and stainless types of free cutting steels. It acts as an alloying agent to improve machine ability of steel castings which are used to manufacture various steel components.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIron(II) sulfide is used as a pigment in hair dyes and ceramics, glasses and bottles, and paint, Also used in lubricants and anodes, as a hydrogen sulfide source, and to treat exhaust gases. And it is also used as resulphurizing agent in the production of carbon, alloy and stainless types of free cutting steels. It acts as an alloying agent to improve machine ability of steel castings which are used to manufacture various steel components.
Solubility
Soluble in water(0.0062g/L ), in acids with evolution of hydrogen sulfide. Insoluble in nitric acid.
Notes
Moisture Sensitive. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agent, acids, air, water/ moisture.
Specifications
| Iron(II) sulfide | |
| 1317-37-9 | |
| FeS | |
| 14,4058 | |
| Soluble in water(0.0062g/L ),in acids with evolution of hydrogen sulfide. Insoluble in nitric acid. | |
| [S--].[Fe++] | |
| 87.91 | |
| 87.91 | |
| Odorless | |
| 4.74 |
| 1195°C | |
| 5000 g | |
| MFCD00011013 | |
| ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide | |
| GNVXPFBEZCSHQZ-UHFFFAOYSA-N | |
| λ²-iron(2+) sulfanediide | |
| 14828 | |
| Technical | |
| Air and moisture sensitive |
Chemical Identifiers
| 1317-37-9 | |
| 87.91 | |
| GNVXPFBEZCSHQZ-UHFFFAOYSA-N | |
| 14828 |
| FeS | |
| MFCD00011013 | |
| ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide | |
| [S--].[Fe++] |
Safety and Handling
EINECSNumber : 215-268-6
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only