missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfide, 99.9%, (trace metal basis), -100 mesh

Catalog No. AC315870100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC315870100 10 g Glass bottle
AC315870500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC315870100 Supplier Thermo Scientific Chemicals Supplier No. 315870100

CAS: 1317-37-9 | FeS | 87.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 1317-37-9
Molecular Formula FeS
Molecular Weight (g/mol) 87.91
MDL Number MFCD00011013
InChI Key GNVXPFBEZCSHQZ-UHFFFAOYSA-N
Synonym ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
PubChem CID 14828
IUPAC Name λ²-iron(2+) sulfanediide
SMILES [S--].[Fe++]

Specifications

Melting Point 1195.0°C
Color Black to Brown or Gray
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9%
Linear Formula FeS
Merck Index 15, 4085
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in acids with the evolution of h2s
Formula Weight 87.92
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 4.84
Density 4.8400g/mL
Chemical Name or Material Iron(II) sulfide, -100 mesh
Show More Show Less
EINECSNumber 215-268-6

RUO – Research Use Only