missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) selenide, 99.9% (metals basis)
Description
Iron(II) selenide is used in electrical semiconductors. It is transformed into superconductor by a purely chemical method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Black |
| Physical Form | Powder |
| Quantity | 50 g |
| Assay Percent Range | 99.9% (Metals basis) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 134.81 |
| Odor | Odorless |
| Sensitivity | Moisture sensitive |
| Chemical Name or Material | Iron(II) selenide |
Safety and Handling
| Hazard Category | H301+H331-H373 |
| Hazard Statement | When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. |
| Precautionary Statement | P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c |
| DOTInformation | DOT Class: 6.1, Packing Group: III |
| EINECSNumber | 215-177-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only