missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) oxalate hydrate, 96%

Catalog No. AA3926136
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3926136 500 g
AA39261A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3926136 Supplier Thermo Scientific Chemicals Supplier No. 03926136

CAS: 516-03-0 | C2FeO4 | 143.86 g/mol

Used as a photographic developer, glass tint, decorative glass colorant, and pigment in plastics, paints, and lacquers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 516-03-0
Molecular Formula C2FeO4
Molecular Weight (g/mol) 143.86
MDL Number MFCD00012475
InChI Key OWZIYWAUNZMLRT-UHFFFAOYSA-L
Synonym ferrous oxalate, iron ii oxalate, unii-dzp4yv3icv, dzp4yv3icv, iron protoxalate, iron, ethanedioato 2--.kappa.o1,.kappa.o2, iron 2+ oxalate, iron oxalate fec2o4, hsdb 463, iron 2+ ; oxalate
PubChem CID 10589
IUPAC Name iron(2+);oxalate
SMILES [Fe++].[O-]C(=O)C([O-])=O

Specifications

Melting Point 190°C (decomposition)
Density 2.28 g/mL
Quantity 500 g
UN Number UN3288
Sensitivity Hygroscopic
Solubility Information Soluble in acids
Formula Weight 143.87 (Anhydrous)
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Iron(II) oxalate hydrate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 208-217-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only