missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) Gluconate Hydrate 95.0+%, TCI America™
$106.25 - $270.29
Chemical Identifiers
| CAS | 22830-45-1 |
|---|---|
| Molecular Formula | C12H26FeO16 |
| Molecular Weight (g/mol) | 482.17 |
| MDL Number | MFCD00150872 |
| InChI Key | OKGNXSFAYMSVNN-SYAJEJNSSA-L |
| Synonym | iron ii gluconate hydrate |
| PubChem CID | 131851469 |
| IUPAC Name | λ²-iron(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) dihydrate |
| SMILES | O.O.[Fe++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O |
Chemical Identifiers
| 22830-45-1 | |
| 482.17 | |
| OKGNXSFAYMSVNN-SYAJEJNSSA-L | |
| 131851469 | |
| O.O.[Fe++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O |
| C12H26FeO16 | |
| MFCD00150872 | |
| iron ii gluconate hydrate | |
| λ²-iron(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) dihydrate |
Specifications
| 22830-45-1 | |
| 25 g | |
| MFCD00150872 | |
| OKGNXSFAYMSVNN-SYAJEJNSSA-L | |
| λ²-iron(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) dihydrate | |
| 131851469 | |
| ≥95.0% (T) |
| Crystalline Powder | |
| C12H26FeO16 | |
| iron ii gluconate hydrate | |
| O.O.[Fe++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O | |
| 482.17 | |
| 446.14 | |
| Iron(II) Gluconate Hydrate |
Safety and Handling
RTECSNumber : LZ5180000
TSCA : No