missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Gluconate Hydrate 95.0+%, TCI America™

Catalog No. G003825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G003825G 25 g
G0038500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G003825G Supplier TCI America Supplier No. G003825G

Chemical Identifiers

CAS 22830-45-1
Molecular Formula C12H26FeO16
Molecular Weight (g/mol) 482.17
MDL Number MFCD00150872
InChI Key OKGNXSFAYMSVNN-SYAJEJNSSA-L
Synonym iron ii gluconate hydrate
PubChem CID 131851469
IUPAC Name λ²-iron(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) dihydrate
SMILES O.O.[Fe++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 446.14
Percent Purity ≥95.0% (T)
Chemical Name or Material Iron(II) Gluconate Hydrate
RTECSNumber LZ5180000
TSCA No