missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) fumarate, 94%

Catalog No. p-7027303
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2395630 250 g
AAB239560B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2395630 Supplier Thermo Scientific Chemicals Supplier No. B2395630
Only null left

CAS: 141-01-5 | C4H2FeO4-2 | 169.901 g/mol

Iron(II) fumarate is used as dietary Supplement, nutrient, iron source in foods and pharmaceuticals, as animal feed additive and in infant formulas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(II) fumarate is used as dietary Supplement, nutrient, iron source in foods and pharmaceuticals, as animal feed additive and in infant formulas.

Solubility
Soluble in water (0.14 g/100 ml at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 141-01-5
Molecular Formula C4H2FeO4-2
Molecular Weight (g/mol) 169.901
MDL Number MFCD00058315
InChI Key OOPLWEDSEDELIX-TYYBGVCCSA-L
Synonym ferrous fumarate, eisenfumarat
PubChem CID 66524104
IUPAC Name (E)-but-2-enedioate;iron
SMILES C(=CC(=O)[O-])C(=O)[O-].[Fe]

Specifications

Melting Point >300°C
Quantity 250 g
Merck Index 14,4046
Solubility Information Soluble in water (0.14g/100 ml at 25°C).
Formula Weight 169.9
Percent Purity 94%
Density 2.44
Chemical Name or Material Iron(II) fumarate
EINECSNumber 205-447-7
RTECSNumber LT1950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only