missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) ethylenediammonium sulfate tetrahydrate, 99%

CAS: 113193-60-5

$100.45 - $211.84

Chemical Identifiers

CAS 113193-60-5
MDL Number MFCD00082474
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3054714
View Documents
Thermo Scientific Chemicals
03054714
25 g
Each for $100.45
Only null left
 
AA3054722
View Documents
Thermo Scientific Chemicals
03054722
100 g
Each for $211.84
Only null left
 
Description

Description

Iron(II) ethylenediammonium sulfate tetrahydrate is used in the preparation of gold nanoparticles by reacting with gold(III)-bis[ethylene-diamine]-chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

113193-60-5
MFCD00082474

Specifications

113193-60-5
FeC2H4(NH3)2(SO4)2·4H2O
25 g
382.14 (310.08 Anhydrous)
Crystalline
Green to White
MFCD00082474
Soluble in water.
99%
Iron(II) ethylenediammonium sulfate tetrahydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 264-357-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only