missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Iron(II) Ethylenediammonium Sulfate Tetrahydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Supplier: Sigma Aldrich 414913100G
Alert:
Warning
Description
Iron(II) Ethylenediammonium Sulfate TetrahydrateSpecifications
| 34962-29-3 | |
| FeSO4 -+ NH3CH2CH2NH3SO4 -+ 4H2O | |
| 100 g | |
| 382.15 g/mol |
| C2H10N2O4S -+ FeSO4 -+ 4H2O | |
| MFCD00149220 | |
| Ferrous ethylenediammonium sulfate tetrahydrate | |
| Respiratory System |