missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) chloride tetrahydrate, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 1 kg |
| Linear Formula | FeCl2 · 4H2O |
| UN Number | UN3260 |
| Formula Weight | 198.81g/mol |
| Percent Purity | ≥99% (RT) |
| Grade | ACS |
| Vapor Pressure | 10 mmHg (693°C) |
| Chemical Name or Material | Iron(II) chloride tetrahydrate |
Safety and Handling
| Hazard Statement | P280-P305 + P351 + P338-P310 |
| RTECSNumber | NO5600000 |